Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rcor3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134985.1 | NP_001128457.1 | 8 - 9 | 1095 | 64 | |
GenBank mRNA | BC169033.1 | - | 8 - 9 | 1095 | 7114893 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |