Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cr1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005265.1 | NP_001005265.1 | 6 - 7 | 1226 | 119 | |
NM_001005330.1 | NP_001005330.1 | 1226 | 119 | |||
NM_019301.2 | NP_062174.1 | 6 - 7 | 1226 | 119 | ||
GenBank mRNA | BC061736.1 | - | 6 - 7 | 995 | 5599318 | 119 |
D42114.1 | - | 6 - 7 | 1035 | 119 | ||
D42115.1 | - | 6 - 7 | 971 | 119 | ||
D42116.1 | - | 3 - 4 | 579 | 119 | ||
FQ220559.1 | - | 2 - 3 | 245 | 119 | ||
L36532.1 | - | 6 - 7 | 994 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |