Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psen2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031087.2 | NP_112349.2 | 4 - 5 | 589 | 118 | |
XM_006250405.2 | XP_006250467.1 | 6 - 7 | 740 | 118 | ||
XM_006250407.2 | XP_006250469.1 | 5 - 6 | 650 | 118 | ||
XM_006250408.2 | XP_006250470.1 | 4 - 5 | 565 | 118 | ||
XM_006250410.2 | XP_006250472.1 | 5 - 6 | 1333 | 118 | ||
XM_008769876.1 | XP_008768098.1 | 5 - 6 | 1332 | 118 | ||
XM_008769877.1 | XP_008768099.1 | 6 - 7 | 880 | 118 | ||
XM_008769878.1 | XP_008768100.1 | 4 - 5 | 1958 | 118 | ||
GenBank mRNA | AB004454.1 | - | 3 - 4 | 499 | 118 | |
BC078805.1 | - | 4 - 5 | 589 | 7109917 | 118 | |
D83700.1 | - | 3 - 4 | 492 | 118 | ||
FQ219614.1 | - | 6 - 7 | 708 | 118 | ||
X99267.1 | - | 4 - 5 | 612 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Rattus norvegicus Probe spans exons |