Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psen2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031087.2 | NP_112349.2 | 6 - 7 | 888 | 79 | |
XM_006250405.2 | XP_006250467.1 | 8 - 9 | 1039 | 79 | ||
XM_006250407.2 | XP_006250469.1 | 7 - 8 | 949 | 79 | ||
XM_006250408.2 | XP_006250470.1 | 6 - 7 | 864 | 79 | ||
XM_006250410.2 | XP_006250472.1 | 7 - 8 | 1632 | 79 | ||
XM_008769876.1 | XP_008768098.1 | 7 - 8 | 1631 | 79 | ||
XM_008769877.1 | XP_008768099.1 | 8 - 9 | 1179 | 79 | ||
XM_008769878.1 | XP_008768100.1 | 6 - 7 | 2257 | 79 | ||
GenBank mRNA | AB004454.1 | - | 5 - 6 | 798 | 79 | |
BC078805.1 | - | 6 - 7 | 888 | 7109917 | 79 | |
FQ219614.1 | - | 8 - 9 | 1009 | 79 | ||
X99267.1 | - | 6 - 7 | 911 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Rattus norvegicus Probe spans exons |