Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tp53bp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_223012.8 | XP_223012.6 | 3 - 4 | 525 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Apoptosis Rattus norvegicus Probe spans exons |