Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tagln2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013127.1 | NP_001013145.1 | 3 - 4 | 412 | 105 | |
XM_006250295.2 | XP_006250357.1 | 3 - 4 | 847 | 105 | ||
GenBank mRNA | BC084703.1 | - | 3 - 4 | 412 | 7300611 | 105 |
FQ221577.1 | - | 3 - 4 | 423 | 105 | ||
FQ223112.1 | - | 3 - 4 | 437 | 105 | ||
FQ228757.1 | - | 3 - 4 | 431 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |