Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smyd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_206851.1 | NP_996733.1 | 10 - 11 | 1150 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |