Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osbpl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013079.1 | NP_001013097.1 | 4 - 5 | 415 | 133 | |
XM_006235816.2 | XP_006235878.1 | 5 - 6 | 664 | 133 | ||
GenBank mRNA | BC091208.1 | - | 4 - 5 | 415 | 7381353 | 133 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |