Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Birc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008762598.1 | XP_008760820.1 | 2 - 3 | 1331 | 58 | |
XM_008762599.1 | XP_008760821.1 | 2 - 3 | 1331 | 58 | ||
XM_008762600.1 | XP_008760822.1 | 2 - 3 | 1331 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Apoptosis 3' Most Detect Genomic DNA Rattus norvegicus Probe spans exons |