Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zkscan5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107119.1 | NP_001100589.1 | 3 - 4 | 1317 | 64 | |
GenBank mRNA | BC167019.1 | - | 3 - 4 | 1233 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |