Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabra4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080587.3 | NP_542154.3 | 3 - 4 | 361 | 127 | |
GenBank mRNA | S55933.1 | - | 3 - 4 | 361 | 127 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Rattus norvegicus Probe spans exons |