Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lphn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130822.1 | NP_570835.1 | 22 - 23 | 3821 | 107 | |
XM_008769968.1 | XP_008768190.1 | 22 - 23 | 3925 | 107 | ||
XM_008769969.1 | XP_008768191.1 | 22 - 23 | 3910 | 107 | ||
XM_008769970.1 | XP_008768192.1 | 22 - 23 | 3925 | 107 | ||
XM_008769971.1 | XP_008768193.1 | 21 - 22 | 3898 | 107 | ||
XM_008769972.1 | XP_008768194.1 | 21 - 22 | 3886 | 107 | ||
XM_008769973.1 | XP_008768195.1 | 21 - 22 | 3898 | 107 | ||
XM_008769974.1 | XP_008768196.1 | 22 - 23 | 3925 | 107 | ||
XM_008769975.1 | XP_008768197.1 | 22 - 23 | 3925 | 107 | ||
XM_008769976.1 | XP_008768198.1 | 21 - 22 | 3898 | 107 | ||
XM_008769977.1 | XP_008768199.1 | 21 - 22 | 3721 | 107 | ||
XM_008769978.1 | XP_008768200.1 | 20 - 21 | 3859 | 107 | ||
XM_008769979.1 | XP_008768201.1 | 20 - 21 | 3694 | 107 | ||
XM_008769980.1 | XP_008768202.1 | 22 - 23 | 3925 | 107 | ||
XM_008769981.1 | XP_008768203.1 | 21 - 22 | 3898 | 107 | ||
XM_008769982.1 | XP_008768204.1 | 22 - 23 | 3924 | 107 | ||
XM_008769983.1 | XP_008768205.1 | 21 - 22 | 3897 | 107 | ||
XM_008769984.1 | XP_008768206.1 | 22 - 23 | 3924 | 107 | ||
XM_008769985.1 | XP_008768207.1 | 20 - 21 | 3693 | 107 | ||
XM_008769986.1 | XP_008768208.1 | 20 - 21 | 3693 | 107 | ||
GenBank mRNA | AF063103.1 | - | 22 - 23 | 3821 | 107 | |
AF081154.1 | - | 20 - 21 | 3615 | 107 | ||
AF081155.1 | - | 20 - 21 | 3615 | 107 | ||
AF081156.1 | - | 20 - 21 | 3615 | 107 | ||
AF081157.1 | - | 21 - 22 | 3819 | 107 | ||
AF081158.1 | - | 21 - 22 | 3819 | 107 | ||
AF081159.1 | - | 21 - 22 | 3819 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Rattus norvegicus Probe spans exons |