Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lphn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008769980.1 | XP_008768202.1 | 24 - 25 | 4145 | 76 | |
XM_008769981.1 | XP_008768203.1 | 23 - 24 | 4118 | 76 | ||
XM_008769985.1 | XP_008768207.1 | 22 - 23 | 3913 | 76 | ||
GenBank mRNA | AF081156.1 | - | 22 - 23 | 3835 | 76 | |
AF081159.1 | - | 23 - 24 | 4039 | 76 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Rattus norvegicus Probe spans exons |