Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnga1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053497.1 | NP_445949.1 | 2 - 3 | 210 | 91 | |
GenBank mRNA | U93851.1 | - | 2 - 3 | 210 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Ion Channel Rattus norvegicus Probe spans exons |