Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Paics | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080910.3 | NP_543186.1 | 6 - 7 | 779 | 65 | |
XM_006250852.2 | XP_006250914.1 | 6 - 7 | 827 | 65 | ||
XM_006250853.2 | XP_006250915.1 | 6 - 7 | 798 | 65 | ||
XM_006250854.2 | XP_006250916.1 | 6 - 7 | 828 | 65 | ||
GenBank mRNA | BC072508.1 | - | 6 - 7 | 769 | 7103198 | 65 |
BC085711.1 | - | 6 - 7 | 779 | 7192781 | 65 | |
D37979.1 | - | 4 - 5 | 684 | 65 |
Gene Symbol: LOC100910308 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751352.3 | XP_003751400.2 | 3 - 4 | 555 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |