Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dclre1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106201.1 | NP_001099671.1 | 4 - 5 | 2470 | 125 | |
XM_006231634.1 | XP_006231696.1 | 5 - 6 | 3146 | 125 | ||
XM_006231635.2 | XP_006231697.1 | 5 - 6 | 3084 | 125 | ||
XM_008760523.1 | XP_008758745.1 | 5 - 6 | 3146 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |