Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxl15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107603.1 | NP_001101073.1 | 3 - 4 | 1005 | 86 | |
GenBank mRNA | FQ235302.1 | - | 3 - 4 | 997 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |