Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nolc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022869.1 | NP_074060.1 | 8 - 9 | 1063 | 67 | |
XM_006231560.2 | XP_006231622.1 | 8 - 9 | 1111 | 67 | ||
GenBank mRNA | M94287.1 | - | 8 - 9 | 1063 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |