Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccng2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105725.2 | NP_001099195.1 | 1 - 2 | 188 | 112 | |
GenBank mRNA | BC169085.1 | - | 1 - 2 | 188 | 7938657 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |