Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccng2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105725.2 | NP_001099195.1 | 3 - 4 | 462 | 113 | |
GenBank mRNA | BC169085.1 | - | 3 - 4 | 462 | 7938657 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |