Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006958.1 | NP_001006959.1 | 15 - 16 | 1340 | 142 | |
XM_006250722.2 | XP_006250784.1 | 15 - 16 | 1410 | 142 | ||
GenBank mRNA | BC083620.1 | - | 15 - 16 | 1340 | 7133993 | 142 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |