Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: G3bp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013989.1 | NP_001014011.1 | 5 - 6 | 822 | 131 | |
XM_006250733.2 | XP_006250795.1 | 5 - 6 | 816 | 131 | ||
XM_006250734.2 | XP_006250796.1 | 5 - 6 | 871 | 131 | ||
XM_006250735.1 | XP_006250797.1 | 5 - 6 | 654 | 131 | ||
XM_006250736.2 | XP_006250798.1 | 4 - 5 | 465 | 131 | ||
GenBank mRNA | BC079225.1 | - | 5 - 6 | 822 | 7132309 | 131 |
FQ212261.1 | - | 5 - 6 | 659 | 131 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |