Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thap6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008770033.1 | XP_008768255.1 | 4 | 67 | ||
XM_008770035.1 | XP_008768257.1 | 4 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus 1700 Both primers and probe map within a single exon |