Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shroom3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100889.1 | NP_001094359.1 | 9 - 10 | 5323 | 87 | |
XM_006250723.2 | XP_006250785.1 | 9 - 10 | 6337 | 87 | ||
XM_006250725.2 | XP_006250787.1 | 7 - 8 | 8110 | 87 | ||
GenBank mRNA | BC091437.1 | - | 1 - 2 | 154 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |