Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Afm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172320.1 | NP_758823.1 | 3 - 4 | 274 | 87 | |
XM_008770001.1 | XP_008768223.1 | 3 - 4 | 322 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |