Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35f4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008770650.1 | XP_008768872.1 | 5 - 6 | 1072 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |