Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013131.1 | NP_001013149.1 | 6 - 7 | 713 | 112 | |
GenBank mRNA | BC091207.1 | - | 6 - 7 | 713 | 7381272 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |