Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Otx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006251810.2 | XP_006251872.1 | 105 | 59 | ||
XM_008770527.1 | XP_008768749.1 | 111 | 59 |
Gene Symbol: LOC100911492 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006251839.2 | XP_006251901.1 | 1 - 2 | 370 | 59 | |
XM_008770622.1 | XP_008768844.1 | 1 - 2 | 370 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |