Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdkn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006251791.2 | XP_006251853.1 | 6 - 7 | 534 | 120 | |
GenBank mRNA | BC167026.1 | - | 5 - 6 | 448 | 8375927 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Protein Phosphatase Mammalian Gene Collection Rattus norvegicus Probe spans exons |