Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cgrrf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053899.1 | NP_446351.1 | 3 - 4 | 514 | 116 | |
GenBank mRNA | FQ218613.1 | - | 3 - 4 | 529 | 116 | |
U66471.1 | - | 3 - 4 | 514 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |