Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006251858.2 | XP_006251920.1 | 1 - 2 | 649 | 127 | |
XM_006251861.2 | XP_006251923.1 | 1 - 2 | 648 | 127 | ||
XM_006251865.2 | XP_006251927.1 | 1 - 2 | 648 | 127 |
Gene Symbol: LOC100911576 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006251450.2 | XP_006251512.1 | 1 - 2 | 647 | 127 | |
XM_006251455.2 | XP_006251517.1 | 1 - 2 | 646 | 127 | ||
GenBank mRNA | AY724499.1 | - | 1 - 2 | 574 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |