Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100509.1 | NP_001093979.1 | 4 - 5 | 309 | 127 | |
GenBank mRNA | BC107950.1 | - | 4 - 5 | 230 | 127 | |
DY570284.1 | - | 4 - 5 | 309 | 127 | ||
FQ213949.1 | - | 4 - 5 | 257 | 127 | ||
FQ214043.1 | - | 4 - 5 | 262 | 127 | ||
FQ227742.1 | - | 4 - 5 | 266 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |