Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ero1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138528.1 | NP_612537.1 | 2 - 3 | 248 | 105 | |
GenBank mRNA | AF489855.1 | - | 2 - 3 | 235 | 105 | |
AY071924.1 | - | 2 - 3 | 248 | 105 | ||
FQ228432.1 | - | 93 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus 1700 Probe spans exons |