Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nsl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109083.1 | NP_001102553.1 | 1 - 2 | 260 | 106 | |
XM_006250488.1 | XP_006250550.1 | 1 - 2 | 253 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |