Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127638.1 | NP_001121110.1 | 5 - 6 | 1094 | 74 | |
XM_008769764.1 | XP_008767986.1 | 5 - 6 | 1237 | 74 | ||
GenBank mRNA | BC166443.1 | - | 5 - 6 | 1094 | 8363035 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Protease Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |