Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Diexf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013986.2 | NP_001014008.1 | 7 - 8 | 1397 | 68 | |
XM_006250482.2 | XP_006250544.1 | 8 - 9 | 1587 | 68 | ||
GenBank mRNA | BC087114.1 | - | 7 - 8 | 1351 | 7193786 | 68 |
FQ217506.1 | - | 8 - 9 | 1570 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |