Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Susd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105982.1 | NP_001099452.1 | 3 - 4 | 538 | 105 | |
XM_006250373.2 | XP_006250435.1 | 3 - 4 | 692 | 105 | ||
XM_006250375.2 | XP_006250437.1 | 3 - 4 | 577 | 105 | ||
XM_008769820.1 | XP_008768042.1 | 3 - 4 | 692 | 105 | ||
XM_008769821.1 | XP_008768043.1 | 3 - 4 | 581 | 105 | ||
XM_008769822.1 | XP_008768044.1 | 3 - 4 | 670 | 105 | ||
XM_008769823.1 | XP_008768045.1 | 3 - 4 | 692 | 105 | ||
GenBank mRNA | FQ212123.1 | - | 3 - 4 | 522 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |