Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snrpb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108592.2 | NP_001102062.1 | 3 - 4 | 287 | 123 | |
XM_006235116.2 | XP_006235178.1 | 3 - 4 | 448 | 123 | ||
GenBank mRNA | BC168760.1 | - | 3 - 4 | 287 | 6888886 | 123 |
FQ213679.1 | - | 3 - 4 | 319 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |