Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcsk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012746.1 | NP_036878.1 | 1 - 2 | 493 | 72 | |
GenBank mRNA | M76706.1 | - | 1 - 2 | 493 | 72 | |
M83746.1 | - | 1 - 2 | 465 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Rattus norvegicus Probe spans exons |