Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcsk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012746.1 | NP_036878.1 | 5 - 6 | 858 | 92 | |
GenBank mRNA | M76706.1 | - | 5 - 6 | 858 | 92 | |
M83746.1 | - | 5 - 6 | 830 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Rattus norvegicus Probe spans exons |