Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polr3f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107784.1 | NP_001101254.1 | 6 - 7 | 781 | 142 | |
XM_008762269.1 | XP_008760491.1 | 6 - 7 | 812 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |