Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gata5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024316.1 | NP_001019487.1 | 4 - 5 | 1239 | 78 | |
XM_008762533.1 | XP_008760755.1 | 4 - 5 | 2704 | 78 | ||
GenBank mRNA | BC085855.1 | - | 4 - 5 | 1239 | 7190097 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |