Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arfgap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145090.3 | NP_659558.1 | 10 - 11 | 1001 | 90 | |
XM_006235734.2 | XP_006235796.1 | 9 - 10 | 1309 | 90 | ||
GenBank mRNA | BC070895.1 | - | 10 - 11 | 982 | 7099840 | 90 |
CV120035.1 | - | 2 - 3 | 92 | 90 | ||
FQ222295.1 | - | 10 - 11 | 1024 | 90 | ||
U35776.1 | - | 10 - 11 | 1001 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |