Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013924.1 | NP_001013946.1 | 3 - 4 | 408 | 89 | |
XM_006235817.2 | XP_006235879.1 | 3 - 4 | 834 | 89 | ||
XM_006235818.2 | XP_006235880.1 | 3 - 4 | 454 | 89 | ||
XM_006235820.2 | XP_006235882.1 | 3 - 4 | 462 | 89 | ||
XM_008762485.1 | XP_008760707.1 | 2 - 3 | 1968 | 89 | ||
XM_008762486.1 | XP_008760708.1 | 3 - 4 | 838 | 89 | ||
XM_008762487.1 | XP_008760709.1 | 4 - 5 | 555 | 89 | ||
XM_008762488.1 | XP_008760710.1 | 4 - 5 | 578 | 89 | ||
XM_008762489.1 | XP_008760711.1 | 3 - 4 | 477 | 89 | ||
XM_008762490.1 | XP_008760712.1 | 4 - 5 | 534 | 89 | ||
XM_008762491.1 | XP_008760713.1 | 3 - 4 | 458 | 89 | ||
XM_008762492.1 | XP_008760714.1 | 2 - 3 | 1968 | 89 | ||
XM_008762493.1 | XP_008760715.1 | 3 - 4 | 485 | 89 | ||
XM_008762494.1 | XP_008760716.1 | 2 - 3 | 1968 | 89 | ||
XM_008762495.1 | XP_008760717.1 | 2 - 3 | 1968 | 89 | ||
GenBank mRNA | BC083707.1 | - | 3 - 4 | 408 | 7190542 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |