Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mid1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022927.1 | NP_075216.1 | 8 - 9 | 1678 | 145 | |
XM_006256825.2 | XP_006256887.1 | 8 - 9 | 2962 | 145 | ||
GenBank mRNA | AF186461.1 | - | 8 - 9 | 1678 | 145 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |