Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glis1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006238561.2 | XP_006238623.1 | 9 - 10 | 2174 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |