Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppcs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039010.2 | NP_001034099.2 | 1 - 2 | 506 | 126 | |
GenBank mRNA | CK474267.1 | - | 1 - 2 | 545 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |