Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171993.1 | NP_741990.1 | 6 - 7 | 1037 | 61 | |
GenBank mRNA | BC085691.1 | - | 6 - 7 | 1021 | 7134739 | 61 |
U05341.1 | - | 6 - 7 | 1037 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |