Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tesk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133396.2 | NP_596887.1 | 7 - 8 | 1063 | 71 | |
XM_006238628.2 | XP_006238690.1 | 7 - 8 | 1294 | 71 | ||
GenBank mRNA | AB049402.1 | - | 7 - 8 | 1064 | 71 | |
BC081737.1 | - | 8 - 9 | 1314 | 71 | ||
BC128727.1 | - | 7 - 8 | 1069 | 7111312 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |