Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001139465.2 | NP_001132937.1 | 2 - 3 | 349 | 88 | |
GenBank mRNA | CK476149.1 | - | 2 - 3 | 354 | 88 | |
FM094601.1 | - | 1 - 2 | 81 | 88 | ||
FQ212504.1 | - | 2 - 3 | 315 | 88 | ||
FQ214069.1 | - | 2 - 3 | 346 | 88 | ||
FQ226828.1 | - | 1 - 2 | 88 | |||
FQ231829.1 | - | 2 - 3 | 333 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |